Please login to your online account to display your discounted pricing

GE Healthcare Dharmacon Accell™ Fluorescent Nontargeting Control siRNA


Provides highly reliable qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake

Manufacturer: ge healthcare dharmacon  D0019500105

 View more versions of this product

Catalog No. D0019500105

  • / Each
 

Description & Specifications

Specifications

Fluorophore FITC
For Use With (Application) Evaluate cytoplamic localization of the dye-labeled Accell™ siRNA FACS analysis to determine general uptake and provides highly reliable qualitative assessment of Accell™ siRNA uptake as determined by Fluorescent micros
Quantity 5nm

Accell Green (FAM)

  • Filter: FITC
  • Absorbance: 494nm
  • Emission Max: 520nm
Accell Red (DY-547)
  • Filter: Cy™3, Rhodamine, or PE
  • Absorbance: 557nm
  • Emission Max: 570nm