Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ Fluorescent Nontargeting Control siRNA
Provides highly reliable qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA FACS analysis to determine general uptake
$258.82 - $258.82
Specifications
For Use With (Application) Evaluate cytoplamic localization of the dye-labeled Accell™ siRNA FACS analysis to determine general uptake and provides highly reliable qualitative assessment of Accell™ siRNA uptake as determined by Fluorescent micros
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
D0019500105
ge healthcare dharmacon
D0019500105
5nm Each for $258.82
Description & Specifications
Specifications
For Use With (Application) Evaluate cytoplamic localization of the dye-labeled Accell™ siRNA FACS analysis to determine general uptake and provides highly reliable qualitative assessment of Accell™ siRNA uptake as determined by Fluorescent micros
Accell Green (FAM)
  • Filter: FITC
  • Absorbance: 494nm
  • Emission Max: 520nm
Accell Red (DY-547)
  • Filter: Cy™3, Rhodamine, or PE
  • Absorbance: 557nm
  • Emission Max: 570nm