Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ Cyclophilin B Control siRNA and Cyclophilin B Pool
Recommended as a positive silencing control
Manufacturer: ge healthcare dharmacon  D0019200120
 View more versions of this product
Catalog No. D0019200120
  • / Each
Description & Specifications
Specifications
Accession Number NM_000942
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Product Type Cyclophilin B Control siRNA—Human
Quantity 20nm
Also known as peptidylprolyl isomerase B (PPIB), this gene is abundantly expressed in most cells and because it is nonessential, knockdown of the corresponding mRNA does not affect cell viability.
  • Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable positive controls for delivery and silencing capability
  • Targets accession numbers: NM_000942 (Human) or NM_011149 (Mouse) or NM_022536 (Rat)