Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon Accell™ Cyclophilin B Control siRNA and Cyclophilin B Pool
Recommended as a positive silencing control
$172.56 - $635.17
Specifications
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
View More Specs
Products
Catalog Number Mfr. No. Accession Number Product Type Quantity Price Quantity    
D0019200105
ge healthcare dharmacon
D0019200105
NM_000942 Cyclophilin B Control siRNA—Human 5nm Each for $172.56
D0019200120
ge healthcare dharmacon
D0019200120
NM_000942 Cyclophilin B Control siRNA—Human 20nm Each for $380.81
D0019200350
ge healthcare dharmacon
D0019200350
NM_022536 Each for $635.17
Description & Specifications
Specifications
For Use With (Application) Designed to aid in the interpretation of experiments with Accell™ siRNA reagents as highly reliable positive controls for delivery and silencing capability
Also known as peptidylprolyl isomerase B (PPIB), this gene is abundantly expressed in most cells and because it is nonessential, knockdown of the corresponding mRNA does not affect cell viability.
  • Designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable positive controls for delivery and silencing capability
  • Targets accession numbers: NM_000942 (Human) or NM_011149 (Mouse) or NM_022536 (Rat)