Please login to your online account to display your discounted pricing

GE Healthcare Dharmacon Accell™ Control siRNA Kits

Accell siRNA represents a breakthrough technology to empower RNAi, without transfection reagents, viral vectors, or instrumentation

$915.82 - $915.82

Specifications

For Use With (Application) Assessing Accell™ technology in a given cell type and to ensure effective controls during initial gene silencing experiments with Accell™ siRNA reagents
View More Specs
Includes:  Five nmol tubes of each of the four validated control siRNAs, 5X siRNA buffer, 100mL delivery media
Products
Catalog Number Mfr. No. Species Product Type Price Quantity    

K005000G102

 
ge healthcare dharmacon
K005000G102
Mouse Accell siRNA Control Kit (Green) Each for $915.82
Description & Specifications

Specifications

For Use With (Application) Assessing Accell™ technology in a given cell type and to ensure effective controls during initial gene silencing experiments with Accell™ siRNA reagents

The control kits are designed to be a starting point for assessing Accell technology in a given cell type, and to ensure effective controls during initial gene silencing experiments with Accell siRNA reagents.

All kits contain:

  • Accell Nontargeting siRNA #1: Distinguish general cellular responses to delivery methods and potential off-target effects from siRNA-mediated sequence-specific silencing
  • Accell Cyclophilin B siRNA and GAPD siRNA: Species-specific siRNAs to provide a positive control for silencing and validation of experimental design and detection methodologies
  • 5X siRNA buffer: The recommended buffer for resuspension of Accell siRNA
  • Accell siRNA delivery media: Specially formulated and optimized for Accell siRNA delivery; required for use with Accell siRNA reagents

Accell siRNA Control Kits (Green) also contain:

  • Accell Green Nontargeting siRNA: FAM-labeled Accell siRNA provides highly reliable qualitative assessment of uptake as determined by fluorescent microscopy to evaluate cytoplasmic localization of the dye-labeled Accell siRNA
    • Filter: FITC
    • Absorbance: 494nm
    • Emission Max: 520nm
Accell siRNA Control Kits (Red) also contain:
  • Accell Red Nontargeting siRNA: Dy-547-labeled Accell siRNA provides highly reliable qualitative assessment of uptake as determined by fluorescent microscopy to evaluate cytoplasmic localization of the dye-labeled Accell siRNA
    • Filter: Cy™3, Rhodamine, or PE
    • Absorbance: 557nm
    • Emission Max: 570nm