Please login to your online account to display your discounted pricing

GE Healthcare CyDye™ DIGE Fluor Minimal Dye Labeling Kits

Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel. Amersham CyDye™ DIGE Fluor Minimal Labeling Kits contain size- and charge-matched fluorescent dyes specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.

$1,118.17 - $2,818.98

Specifications

Color Blue, Green, Red
Format Solid
Storage Requirements -15° to -30°C
View More Specs
Products
Catalog Number Mfr. No. Product Type Quantity Price Quantity    

45-002-245

 
ge healthcare
28934530
Amersham CyDye DIGE Fluor Minimal Dye Labeling Kit 2nmol of each: Cy2, Cy3, and Cy5 Each for $1,118.17

36-102-0374

 
ge healthcare
25801065
Amersham CyDye DIGE Fluor Minimal Labeling Kit 5nmol of each: Cy2, Cy3, and Cy5 Each for $2,818.98
Description & Specifications

Specifications

Color Blue, Green, Red
Format Solid
Storage Requirements -15° to -30°C

  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy

All minimal dyes allow the labeling of 50μg protein per labeling reaction. Minimal Dye Labeling Kits contain 2nmol or 5nmol of each of the minimal dyes.

For general 2-D applications where sufficient amounts of sample are available.