Please login to your online account to display your discounted pricing
GE Healthcare Amersham™ TE 77 and TE 77 PWR Semi-Dry Transfer Unit
Electrotransfer proteins from polyacrylamide gels up to 14 x 16cm (TE 77 PWR) and 21 x 26cm (TE 77) in size in less than one hour
$1,862.24 - $1,862.24
Specifications
Product Type Semi Dry Transfer Unit
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    
45-001-782
ge healthcare
11001341
25 Sheets of blotting Paper, 50 sheets of cellophane, two solid masks and one mask each for 14 x 16cm and 7 x 8cm gels Each for $1,862.24
Description & Specifications
Specifications
Product Type Semi Dry Transfer Unit
The TE 77 and TE 77 PWR offers the convenience of a built-in power supply, and a unique automatic stopping feature. The transfer is stopped when buffer gets depleted, preventing overheating.
  • Long-lasting, perforated platinum-titanium and stainless-steel electrodes give uniform, contamination-free transfers
  • Requires minimal buffer, only enough to saturate blotting papers and membranes
  • Uses low current (0.5A) and voltage (30V) for transfers: safer and more economical
  • Suitable for gels up to 14 × 16cm and 21 × 26cm
  • Tested for use with Amersham™ ECL Western Blotting reagents