GE Healthcare Amersham™ Rediprime II DNA Labeling System
Long shelf-life and simple, fast protocol. Amersham™ Rediprime II DNA Labeling System consists of premixed, individually dispensed, random-prime reactions, freeze-dried for ambient stability.
$585.00 - $585.00
Specifications
For Use With (Application) Labeling of DNA from a variety of sources to a high specific activity for use in southern and northern blot hybridizations
View More Specs
Includes: Each system includes the following components, in quantities sufficient for 30 or 60 reactions (dependent on product ordered): Premixed, predispensed tubes, 30X labeling mix, and control DNA.
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
45-000-812 GE Healthcare
RPN1633
30 Reactions Each for $585.00
Specifications
Description
Simply add substrate and [α-32P]dCTP. As little as 25ng substrate DNA can be used.
  • Ambient temperature-stable reactions with a color check for the labeling of DNA to specific activities of >109dpm/μg
  • Long shelf life and simple, fast protocol; labeling reaction complete in 10 min.
  • No need to remove unincorporated nucleotides before hybridization
  • DNA can be labeled in solution or in the presence of low melting point agarose
Suitable for use with dCTP only.
Description & Specifications
Specifications
Labeling of DNA from a variety of sources to a high specific activity for use in southern and northern blot hybridizations