GE Healthcare Amersham™ Rapid-hyb Buffer
Pre-mixed, rate-enhancing hybridization buffer. Amersham™ Rapid-hyb Buffer reduces hybridization time for detecting single-copy mammalian genes to just two hours.
$217.00 - $585.27
Specifications
For Use With (Application) Radiolabeled DNA, RNA or Oligonucleotide probes
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
45-000-814 GE Healthcare
RPN1636
500mL Each for $585.27
45-000-813 GE Healthcare
RPN1635
125mL Each for $217.00
Specifications
Description
  • For use with radiolabeled DNA, RNA, or oligonucleotide probes
  • Pre-mixed solution improves reproducibility between experiments
  • Suited for Southern and Northern blotting applications
  • Available in two different quantities, sufficient for 1000cm2 membrane and 4000cm2 membrane
Developed for use with nylon membranes; not suitable for use with PVDF or nitrocellulose membranes.
Description & Specifications
Specifications
Radiolabeled DNA, RNA or Oligonucleotide probes