GE Healthcare Amersham™ Hyperscreen™ Intensifying Screen
Offers rapid results and high sensitivity for detecting 32P and 125I in Southern, Northern, and Western blots
$360.00 - $360.00
Specifications
Product Type Autoradiography Screen
For Use With (Application) Used with pre-flashed film to give an increase in sensitivity while retaining a linear response that allows quantitation of results
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Length (Metric) Width (Metric) Length (English) Width (English) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Length (English) Width (English) Price Quantity  
45-000-820 GE Healthcare
RPN1669
203mm 254mm 7.99 in. 10 in. Each for $360.00
Specifications
Description
  • Light emission from calcium tungstate phosphor is fully compatible with Hyperfilm™ MP
  • Use with Sensitize™ Pre-Flash Unit for even higher sensitivity and accurate quantitation
  • Fine grain screens give clear band resolution
  • Available in a full range of sizes
Description & Specifications
Specifications
Autoradiography Screen
Used with pre-flashed film to give an increase in sensitivity while retaining a linear response that allows quantitation of results