Cytiva (Formerly GE Healthcare Life Sciences) Amersham™ Hybond™ -XL Membranes
Use for optimal signal-to-noise ratios when using radioactively labeled probes. Cytiva Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.
$455.21 - $527.63
Specifications
Binding Capacity Up to 600μg/cm2
For Use With (Equipment) Radioactive probes
Material Nylon, Highly Positively Charged
View More Specs
Products 5
Catalog Number Mfr. No. Length (Metric) Width (Metric) Format Quantity Price Quantity  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Format Quantity Price Quantity  
45-001-151
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
RPN203S
20 cm 3 m Roll 1 Roll Each for $509.93
45-001-148
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
RPN303S
30 cm 3 m Roll 1 Roll Each for $527.63
45-001-149
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
RPN1520S
15 cm 20 cm Sheet 10 Sheets Each for $455.21
45-001-147
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
RPN2020S
20 cm 20 cm Sheet 10 Sheets Each for $492.77
45-001-152
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
RPN2222S
22.2 cm 22.2 cm Sheet 10 Sheets Each for $490.51
Description
Description
  • Greater concentration and more even distribution of positive charge than other charged nylon membranes
  • Binding capacity for nucleic acids up to 600μg/cm2
  • Efficient binding of small fragments
  • Effective probe removal
  • Suitable for reprobing
  • Discs are also available
Specifications
Specifications
Up to 600μg/cm2
Radioactive probes
Nylon, Highly Positively Charged