Cytiva (Formerly GE Healthcare Life Sciences) Amersham™ Hybond™ -XL Membranes
Use for optimal signal-to-noise ratios when using radioactively labeled probes. Cytiva Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) RPN203S
 View more versions of this product
Catalog No. 45-001-151
Description
Description
  • Greater concentration and more even distribution of positive charge than other charged nylon membranes
  • Binding capacity for nucleic acids up to 600μg/cm2
  • Efficient binding of small fragments
  • Effective probe removal
  • Suitable for reprobing
  • Discs are also available
Specifications
Specifications
1 Roll
Nylon, Highly Positively Charged
3 m
Up to 600μg/cm2
Roll
20 cm
Radioactive probes
Documents