missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GE Healthcare Amersham™ Hybond™ -NX Membranes

Neutral nylon membrane developed specifically for use with low hybridization buffer volumes and high-throughput applications

$485.50 - $502.15

Specifications

Binding Capacity Up to 600μg/cm2
Width (Metric) 3m
Format Roll
For Use With (Equipment) Radioactive probes
Material Nylon, Highly Positively Charged
View More Specs

Products 2
Catalog Number Mfr. No. Length (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Price Quantity  
45-000-855 View Documents GE Healthcare
RPN203T
20cm Each for $485.50
 
45-000-933 View Documents GE Healthcare
RPN303T
30cm Each for $502.15
 
Description

Description

  • Binding capacity for nucleic acids 600μg/cm2
  • High tensile strength withstands multiple reprobing

Specifications

Specifications

Up to 600μg/cm2
Roll
Nylon, Highly Positively Charged
3m
Radioactive probes