Cytiva (Formerly GE Healthcare Life Sciences) Amersham™ Hybond™-N Membranes
Use for high sensitivity applications in DNA and RNA blotting. Cytiva Life Sciences™Amersham™ Hybond™-N Membranes is a strong supported membrane that is inherently hydrophilic and requires no pre-wetting.
$249.72 - $536.67
Specifications
Binding Capacity Up to 600μg/cm2
Material Nylon, Neutral
View More Specs
Products 4
Catalog Number Mfr. No. Description Format Price Quantity  
Catalog Number Mfr. No. Description Format Price Quantity  
45-000-854
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
RPN203N
20cm x 3m Roll Each for $504.25
45-000-932
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
RPN303N
30cm x 3m Roll Each for $536.67
45-000-842
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
RPN2020N
20 x 20cm Sheet Each for $374.15
45-000-953
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
RPN82N
8.2cm Disc Each for $249.72
Description
Description
  • Binding capacity for nucleic acids up to 600μm/cm2
  • Inherently hydrophilic and requires no prewetting
  • Nucleic acids can be crosslinked using UV light
Specifications
Specifications
Up to 600μg/cm2
Nylon, Neutral