Please login to your online account to display your discounted pricing

GE Healthcare Amersham™ ECL™ Precast Gels

Use precast gels to save time and obtain high quality results. Amersham™ ECL™ Precast Gels are convenient to handle and perfect for downstream detection steps.

$125.93 - $125.93

Specifications

Buffer Tris HCl
Color Clear
Width (Metric) Gel 8cm
For Use With (Application) Polyacrylamide Gel electrophoresis
Height (Metric) 7.5cm
View More Specs
Products
Catalog Number Mfr. No. Concentration Format Price Quantity    

45-002-462

 
ge healthcare
28989805
12% 10-well Each for $125.93

45-002-463

 
ge healthcare
28989806
4 to 12% 10-well Each for $125.93
Description & Specifications

Specifications

Buffer Tris HCl
Color Clear
Width (Metric) Gel 8cm
For Use With (Application) Polyacrylamide Gel electrophoresis
Height (Metric) 7.5cm
Shelf Life 12 months
Storage Requirements 4° to 8°C
Thickness (Metric) 0.14cm
Width (English) Gel 3.15 in.

Homogeneous and gradient gels for horizontal protein electrophoresis.

  • Amersham™ ECL™ Gels have a shelf life of up to 12 months with no compromise in performance over time
  • Gels are available in 15 and 10 well formats as either homogenous (10 and 12%) or gradient (4-12%, 8-16% and 4-20%)
  • Compatible with standard Laemmli buffers
  • Works well with standard protein detection protocols
  • Amersham ECL Gel is cast without SDS, allowing the user to define the separation conditions by the composition of the running buffer and sample loading buffer