Please login to your online account to display your discounted pricing

GE Healthcare Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)

Suitable for general labeling of amine residues

$1,179.24 - $7,564.00

Specifications

For Use With (Application) Suitable for general labeling of Amine residues
Includes Foil packs, each containing 5, 10 or 50mg dried dye
Storage Requirements 2° to 8°C
View More Specs
Products
Catalog Number Mfr. No. Product Type Quantity Price Quantity    

45-000-693

 
ge healthcare
PA12000
Cy2 bis-NHS Ester 5mg Each for $1,179.24

45-000-695

 
ge healthcare
PA13000
Cy3 bis-NHS Ester 5mg Each for $1,217.68

45-000-697

 
ge healthcare
PA13004
Cy3 bis-NHS Ester 10mg Each for $1,764.52

45-000-702

 
ge healthcare
PA15002
Cy5 bis-NHS Ester 50mg Each for $7,205.64

45-000-703

 
ge healthcare
PA15004
Cy5 bis-NHS Ester 10mg Each for $1,764.52

45-000-707

 
ge healthcare
PA15500
Cy5.5 bis-NHS Ester 5mg Each for $1,227.60

45-000-708

 
ge healthcare
PA15502
Cy5.5 bis-NHS Ester 50mg Each for $7,564.00

45-000-712

 
ge healthcare
PA17004
Cy7 bis-NHS Ester 10mg Each for $1,796.00

45-000-701

 
ge healthcare
PA15000
Cy5 bis-NHS Ester 5mg Each for $1,227.60
Description & Specifications

Specifications

For Use With (Application) Suitable for general labeling of Amine residues
Includes Foil packs, each containing 5, 10 or 50mg dried dye
Storage Requirements 2° to 8°C

Large range of bulk packs available for greatest economy. The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.