missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GE Healthcare Amersham™ CyDye™ Post-Labeling Reactive Dye Packs

Reactive CyDye fluors specifically optimized for nucleic acid microarray labeling. GE Healthcare CyDye Post-Labelling Reactive Dye Pack provides conveniently packaged, highly reactive Cy3 and Cy5 monofunctional NHS ester to label probes via the post-labeling (amino allyl) route.

$448.00 - $448.00

Specifications

Quantity 12 Vials
For Use With (Application) Only reactive CyDye fluors specifically optimized for Nucleic Acid microarray labeling
Storage Requirements -15° to -30°C
Sufficient For One labeling reaction of up to 0.5 to 2μg of aa-cDNA or 3 to 10μg aa-RNA
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type pH Range Price Quantity  
Catalog Number Mfr. No. Includes Product Type pH Range Price Quantity  
45-001-270 GE Healthcare
RPN5661
12 Vials Cy3 NHS Ester and 12 vials Cy5 NHS Ester CyDye Post-Labeling Reactive Dye Pack 8.5 to 9.5 Each for $448.00
Specifications

Description

  • High reactivity Cy™3 and Cy™5 dyes: 40,000pmol reactive dye per vial, at >75% reactive dye content
  • Each vial is sufficient for one labeling reaction of up to 0.5-2μg of aa-cDNA or 3-10μg aa-RNA
  • Ready-to-use, individually dispensed

Description & Specifications

Specifications

12 Vials
Only reactive CyDye fluors specifically optimized for Nucleic Acid microarray labeling
-15° to -30°C
One labeling reaction of up to 0.5 to 2μg of aa-cDNA or 3 to 10μg aa-RNA