GE Healthcare Amersham™ CDP-Star Detection Reagents
Rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase.  GE Healthcare Amersham™ CDP-Star Detection Reagents label DNA or RNA probes during a 30-minute reaction and resulting probes can be used without purification.
$452.61 - $452.61
Specifications
For Use With (Application) Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
Includes Labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star detection reagents
Quantity Each
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Sufficient For Price Quantity  
Catalog Number Mfr. No. Sufficient For Price Quantity  
45-000-937 GE Healthcare
RPN3682
2500cm2 membrane Each for $452.61
Specifications
Description
  • Hybridization stringency can be controlled by elevating temperatures up to 75°C
  • Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
  • Rapid protocol saves two to three hours over conventional indirect methods
Order Info
Each system include the following reagents, sufficient for 25 or 50 labeling reactions and detection of 2500cm2 or 5000cm2 membrane (depending on product ordered): labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star detection reagents.
Description & Specifications
Specifications
Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
Labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star detection reagents
Each