Please login to your online account to display your discounted pricing
GE Healthcare Accessories and Replacement Parts for Amersham™ VacuGene XL Vacuum Blotting System
VacuGene XL vacuum blotting system is the choice for nucleic acid blotting.
$101.33 - $392.94
Specifications
For Use With (Equipment) Amersham™ VacuGene™ XL vacuum blotting system
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
45-000-419
ge healthcare
18101043
Gel Supporting Screen Each for $105.49
45-000-422
ge healthcare biosciences
18101096
Liquid Trap Each for $392.94
45-500-113
ge healthcare
18101044
Each for $101.33
Description & Specifications
Specifications
For Use With (Equipment) Amersham™ VacuGene™ XL vacuum blotting system
  • Rapidly transfer DNA from agarose gels up to 20 x 30cm in less than 1 hour
  • Efficient and reproducible transfers improve blotting results
  • Carry out all pretransfer steps in the unit
  • Precise vacuum control for optimal transfer conditions
  • RNA transfers in less than 4 hours