GE Cameo™ PTFE Syringe Filters without Glass Prefilters
For use with aggressive acids, bases and for degassing solvents
Manufacturer:  Maine Manufacturing MM1224151
 View more versions of this product
Catalog No. DDF04T30NM
Specifications Description & Specifications
Specifications
Cameo 30F Syringe Filter; 120mL
TefSep PTFE
4.8cm2
30mm
0.45μm
200/Pk.