GE Cameo™ PTFE Syringe Filters without Glass Prefilters
For use with aggressive acids, bases and for degassing solvents
$145.96 - $1,081.36
Specifications
Material TefSep PTFE
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Pore Size Quantity Diameter (Metric) Type Price Quantity  
Catalog Number Mfr. No. Pore Size Quantity Diameter (Metric) Type Price Quantity  
DDF02T3050 Maine Manufacturing
MM1224143
0.22μm 50/Pk. 30mm Cameo 30F Syringe Filter; 120mL Pack of 50 Each for $298.32
DDF02T1750 Maine Manufacturing
MM1224780
0.22μm 50/Pk. 17mm Cameo 17F Syringe Filter; 12mL Pack of 50 Each for $145.96
DDF02T17NB Maine Manufacturing
MM1224782
0.22μm 500/Pk. 17mm Cameo 17F Syringe Filter;12mL Pack of 500 Each for $1,081.36
DDF02T17NM Maine Manufacturing
MM1224781
0.22μm 200/Pk. 17mm Cameo 17F Syringe Filter; 12mL Pack of 200 Each for $494.51
DDF02T30NM Maine Manufacturing
MM1224144
0.22μm 200/Pk. 30mm Cameo 30F Syringe Filter; 120mL Pack of 200 Each for $813.25
DDF04T1750 Maine Manufacturing
MM1224787
0.45μm 50/Pk. 17mm Cameo 17F Syringe Filter; 12mL Pack of 50 Each for $155.51
DDF04T17NB Maine Manufacturing
MM1224789
0.45μm 500/Pk. 17mm Cameo 17F Syringe Filter; 12mL Pack of 500 Each for $1,081.36
DDF04T17NM Maine Manufacturing
MM1224788
0.45μm 200/Pk. 17mm Cameo 17F Syringe Filter; 12mL Pack of 200 Each for $494.51
DDF04T3050 Maine Manufacturing
MM1224150
0.45μm 50/Pk. 30mm Cameo 30F Syringe Filter; 120mL Pack of 50 Each for $280.02
DDF04T30NM Maine Manufacturing
MM1224151
0.45μm 200/Pk. 30mm Cameo 30F Syringe Filter; 120mL Pack of 200 Each for $860.62
Specifications
Description
  • Naturally hydrophobic TefSep PTFE membrane filter will not release extractables into sample
  • Cameo 17F. For HPLC solvent and gas chromatography
  • Cameo 30F. For gas chromatography
  • Sterilizes or clarifies aggressive solvents, acids and bases
  • Use with vent filters to sterilize air, gases or aerosols
Manufactured by GE Infrastructure Water & Process Technologies.
Description & Specifications
Specifications
TefSep PTFE