Please login to your online account to display your discounted pricing

GBR 13069 dihydrochloride, Tocris

Potent dopamine uptake inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material GBR 13069 dihydrochloride
Assay Percent Range >98%
Molecular Formula C28H30F2N2O·2HCl
Formula Weight 521.48
Synonym (1-(2-(Bis-(4-fluorophenyl)methoxy)ethyl)-4-(3-phenyl-2-propenyl)piperazine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

042050

 
r & d systems
042050
50mg Each for $682.67

042010

 
r & d systems
042010
10mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 521.48
Assay Percent Range >98%
Chemical Name or Material GBR 13069 dihydrochloride
Synonym (1-(2-(Bis-(4-fluorophenyl)methoxy)ethyl)-4-(3-phenyl-2-propenyl)piperazine dihydrochloride
CAS 67469-45-8
Molecular Formula C28H30F2N2O·2HCl
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature