Please login to your online account to display your discounted pricing

GBR 13069 dihydrochloride, Tocris


Potent dopamine uptake inhibitor

Quantity 50mg

Manufacturer: r & d systems  042050

 View more versions of this product

Catalog No. 042050

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 521.48
Assay Percent Range >98%
Chemical Name or Material GBR 13069 dihydrochloride
Synonym (1-(2-(Bis-(4-fluorophenyl)methoxy)ethyl)-4-(3-phenyl-2-propenyl)piperazine dihydrochloride
CAS 67469-45-8
Molecular Formula C28H30F2N2O·2HCl
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material GBR 13069 dihydrochloride
Assay Percent Range >98%
Molecular Formula C28H30F2N2O·2HCl
Formula Weight 521.48
Synonym (1-(2-(Bis-(4-fluorophenyl)methoxy)ethyl)-4-(3-phenyl-2-propenyl)piperazine dihydrochloride
Accessories

Desiccate at Room Temperature