Please login to your online account to display your discounted pricing

GBR 12935 dihydrochloride, Tocris

Selective dopamine uptake inhibitor

$88.05 - $323.08

Chemical Identifiers

Chemical Name or Material GBR 12935 dihydrochloride
Assay Percent Range >98%
Molecular Formula C28H34N2O·2HCl
Formula Weight 487.51
Synonym 1-(2-Diphenylmethoxyethyl)-4-(3-phenylpropyl)piperazine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

051410

 
r & d systems
051410
10mg Each for $88.05

051450

 
r & d systems
051450
50mg Each for $323.08
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 487.51
CAS 67469-81-2
Molecular Formula C28H34N2O·2HCl
Assay Percent Range >98%
Chemical Name or Material GBR 12935 dihydrochloride
Synonym 1-(2-Diphenylmethoxyethyl)-4-(3-phenylpropyl)piperazine dihydrochloride
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C