Please login to your online account to display your discounted pricing

GBR 12935 dihydrochloride, Tocris


Selective dopamine uptake inhibitor

Quantity 50mg

Manufacturer: r & d systems  051450

 View more versions of this product

Catalog No. 051450

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 487.51
CAS 67469-81-2
Molecular Formula C28H34N2O·2HCl
Assay Percent Range >98%
Chemical Name or Material GBR 12935 dihydrochloride
Synonym 1-(2-Diphenylmethoxyethyl)-4-(3-phenylpropyl)piperazine dihydrochloride
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material GBR 12935 dihydrochloride
Assay Percent Range >98%
Molecular Formula C28H34N2O·2HCl
Formula Weight 487.51
Synonym 1-(2-Diphenylmethoxyethyl)-4-(3-phenylpropyl)piperazine dihydrochloride
Accessories

Desiccate at -20°C