missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ GBR 12935 dihydrochloride

A very potent and selective inhibitor of dopamine uptake

$127.08 - $439.50

Chemical Identifiers

CAS 67469-81-2
Molecular Formula C28H36Cl2N2O
Molecular Weight (g/mol) 487.509
MDL Number MFCD00083175
InChI Key NQWRSILGEXNJIT-UHFFFAOYSA-N
Synonym gbr 12935 dihydrochloride, gbr-12935 dihydrochloride, gbr 12935 dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxyethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-benzhydryloxyethyl-4-3-phenylpropyl piperazine;dihydrochloride, 1-2-benzhydryloxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl-piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazinediium dichloride, gbr12935 dihydrochloride
PubChem CID 11957553
ChEBI CHEBI:64091
IUPAC Name 1-(2-benzhydryloxyethyl)-4-(3-phenylpropyl)piperazine;dihydrochloride
SMILES C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62991MA View Documents Alfa Aesar™
J62991MA
10mg Each for $127.08
 
AAJ62991MF View Documents Alfa Aesar™
J62991MF
50mg Each for $439.50
 
Specifications

Chemical Identifiers

67469-81-2
487.509
NQWRSILGEXNJIT-UHFFFAOYSA-N
11957553
1-(2-benzhydryloxyethyl)-4-(3-phenylpropyl)piperazine;dihydrochloride
C28H36Cl2N2O
MFCD00083175
gbr 12935 dihydrochloride, gbr-12935 dihydrochloride, gbr 12935 dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxyethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-benzhydryloxyethyl-4-3-phenylpropyl piperazine;dihydrochloride, 1-2-benzhydryloxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl-piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazinediium dichloride, gbr12935 dihydrochloride
CHEBI:64091
C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl

Specifications

White
67469-81-2
MFCD00083175
NQWRSILGEXNJIT-UHFFFAOYSA-N
1-(2-benzhydryloxyethyl)-4-(3-phenylpropyl)piperazine;dihydrochloride
11957553
487.51
GBR 12935 dihydrochloride
10mg
C28H36Cl2N2O
gbr 12935 dihydrochloride, gbr-12935 dihydrochloride, gbr 12935 dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxyethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-benzhydryloxyethyl-4-3-phenylpropyl piperazine;dihydrochloride, 1-2-benzhydryloxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl-piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazinediium dichloride, gbr12935 dihydrochloride
C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl
487.509
CHEBI:64091
Powder
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C