missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ GBR 12909 dihydrochloride

Dopamine uptake inhibitor

$160.38 - $727.88

Chemical Identifiers

CAS 67469-78-7
Molecular Formula C28H34Cl2F2N2O
Molecular Weight (g/mol) 523.49
MDL Number MFCD00055193
InChI Key MIBSKSYCRFWIRU-UHFFFAOYSA-N
Synonym vanoxerine dihydrochloride, gbr 12909 dihydrochloride, vanoxeamine, gbr-12909 dihydrochloride, vanoxerine hydrochloride, unii-mwo1ip03ev, mwo1ip03ev, vanoxerine dihydrochloride gbr12909, gbr12909 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride
PubChem CID 104920
ChEBI CHEBI:64086
IUPAC Name 1-[2-[bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine;dihydrochloride
SMILES C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60684MA View Documents Alfa Aesar™
J60684MA
10mg Each for $160.38
 
AAJ60684MF View Documents Alfa Aesar™
J60684MF
50mg Each for $727.88
 
Specifications

Chemical Identifiers

67469-78-7
523.49
MIBSKSYCRFWIRU-UHFFFAOYSA-N
104920
1-[2-[bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine;dihydrochloride
C28H34Cl2F2N2O
MFCD00055193
vanoxerine dihydrochloride, gbr 12909 dihydrochloride, vanoxeamine, gbr-12909 dihydrochloride, vanoxerine hydrochloride, unii-mwo1ip03ev, mwo1ip03ev, vanoxerine dihydrochloride gbr12909, gbr12909 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride
CHEBI:64086
C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.Cl.Cl

Specifications

White
67469-78-7
MFCD00055193
Soluble in DMSO to 50mM; Soluble to 5mM in water with warming
C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.Cl.Cl
523.49
CHEBI:64086
Powder
10mg
C28H34Cl2F2N2O
vanoxerine dihydrochloride, gbr 12909 dihydrochloride, vanoxeamine, gbr-12909 dihydrochloride, vanoxerine hydrochloride, unii-mwo1ip03ev, mwo1ip03ev, vanoxerine dihydrochloride gbr12909, gbr12909 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride
MIBSKSYCRFWIRU-UHFFFAOYSA-N
1-[2-[bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine;dihydrochloride
104920
523.49
GBR 12909 dihydrochloride