Please login to your online account to display your discounted pricing

GBR 12783 dihydrochloride, Tocris


Potent, selective dopamine uptake inhibitor

Quantity 10mg

Manufacturer: r & d systems  051310

 View more versions of this product

Catalog No. 051310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 485.5
Chemical Name or Material GBR 12783 dihydrochloride
Molecular Formula C28H32N2O·2HCl
Recommended Storage Desiccate at -20°C
Assay Percent Range >99%
Synonym 1-(2-Diphenylmethoxyethyl)-4-(3-phenyl-2-propenyl)-piperazine dihydrochloride
CAS 67469-75-4

Specifications

Chemical Name or Material GBR 12783 dihydrochloride
Assay Percent Range >99%
Molecular Formula C28H32N2O·2HCl
Formula Weight 485.5
Synonym 1-(2-Diphenylmethoxyethyl)-4-(3-phenyl-2-propenyl)-piperazine dihydrochloride
Accessories

Desiccate at -20°C