missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gatifloxacin sesquihydrate, Thermo Scientific Chemicals
A broad spectrum antibiotic
$59.42 - $148.56
Chemical Identifiers
CAS | 180200-66-2 |
---|---|
Molecular Formula | C19H22FN3O4 |
Molecular Weight (g/mol) | 375.40 |
MDL Number | MFCD00895399 |
InChI Key | XUBOMFCQGDBHNK-UHFFFAOYNA-N |
PubChem CID | 5379 |
ChEBI | CHEBI:5280 |
IUPAC Name | 1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid |
SMILES | COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6603603
|
Thermo Scientific Chemicals
J6603603 |
1 g |
Each for $59.42
|
|
|||||
AAJ6603606
|
Thermo Scientific Chemicals
J6603606 |
5 g |
Each for $148.56
|
|
|||||
Description
A broad spectrum antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
180200-66-2 | |
375.40 | |
XUBOMFCQGDBHNK-UHFFFAOYNA-N | |
CHEBI:5280 | |
COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1 |
C19H22FN3O4 | |
MFCD00895399 | |
5379 | |
1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid |
Specifications
Gatifloxacin Sesquihydrate | |
180200-66-2 | |
MFCD00895399 | |
Soluble in 1N NaOH at 10mg/mL | |
COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1 | |
375.40 | |
CHEBI:5280 | |
White to Yellow | |
684°C |
Powder | |
C19H22FN3O4 | |
14,4376 | |
XUBOMFCQGDBHNK-UHFFFAOYNA-N | |
1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid | |
5379 | |
402.42 | |
1 g | |
Gatifloxacin sesquihydrate |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
RTECSNumber : VB1983600
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only