Please login to your online account to display your discounted pricing

Alfa Aesar™ Gastrin Releasing Peptide, Porcine

For cellular and molecular biology applications

Specifications

Product Type Gastrin Releasing Peptide, porcine
Cross Reactivity Porcine
Molecular Weight 2805.40
Storage Requirements -30° to -10°C
Sequence Ala-Pro-Val-Ser-Val-Gly-Gly-Gly-Thr-Val-Leu-Ala-Lys-Met-Tyr-Pro-Arg-Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66527EXD

 
alfa aesar™
J66527EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66527MCR

 
alfa aesar™
J66527MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Gastrin Releasing Peptide, porcine
Cross Reactivity Porcine
Molecular Weight 2805.40
Storage Requirements -30° to -10°C
Sequence Ala-Pro-Val-Ser-Val-Gly-Gly-Gly-Thr-Val-Leu-Ala-Lys-Met-Tyr-Pro-Arg-Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC