Please login to your online account to display your discounted pricing

Alfa Aesar™ Gastric Inhibitory Peptide, Porcine

For cellular and molecular biology applications

Specifications

Product Type Gastric Inhibitory Peptide, porcine
Cross Reactivity Porcine
Molecular Weight 4975.70
Storage Requirements -30° to -10°C
Sequence GIP, porcine
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66401EXD

 
alfa aesar™
J66401EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66401MCR

 
alfa aesar™
J66401MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Gastric Inhibitory Peptide, porcine
Cross Reactivity Porcine
Molecular Weight 4975.70
Storage Requirements -30° to -10°C
Sequence GIP, porcine
Format Solid
Regulatory Status RUO
Purification Method HPLC