missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gastric Inhibitory Peptide (1-30), Porcine

For cellular and molecular biology applications

$804.41 - $1,229.59

Specifications

Product Type Gastric Inhibitory Peptide (1-30), porcine
Cross Reactivity Porcine
Molecular Weight 3552.10
Storage Requirements -30° to -10°C
Sequence Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-Arg-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66088LB0
View Documents
Alfa Aesar™
J66088LB0
5mg Each for $804.41
 
AAJ66088MA
View Documents
Alfa Aesar™
J66088MA
10mg Each for $1,229.59
 
Specifications

Specifications

Gastric Inhibitory Peptide (1-30), porcine
3552.10
Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-Arg-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys
RUO
Porcine
-30° to -10°C
Solid
HPLC

For Research Use Only