missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GARP Rat anti-Mouse, PerCP-eFluor 710, Clone: YGIC86, eBioscience™

Rat Monoclonal Antibody

$183.48 - $473.88

Specifications

Antigen GARP
Clone YGIC86
Host Species Rat
Gene Alias Lrrc32, Garpin
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016113 Life Technologies
LS46989180
25μg Each for $183.48
5016114 Life Technologies
LS46989182
100μg Each for $473.88
Specifications

Description

The YGIC86 monoclonal antibody reacts with mouse Glycoprotein A Repetitions Predominant (GARP, also known as Lrrc32 or Garpin). GARP is an approximately 80kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on Treg cells has been reported to be necessary for the suppressive function of Treg cells, possibly related to its role as a cell surface receptor for LAP/TGF beta.

When looking for expression of GARP on platelets, it is recommended to use the Foxp3 Staining Buffer Set (cat. 00-5523) to fix the platelets before staining with this YGIC86 monoclonal antibody. Fixation/permeabilization is not necessary when staining Treg cells.

YGIC86 does not cross-react to rat GARP.
Description & Specifications

Specifications

GARP
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
YGIC86
Lrrc32, Garpin
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL