missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GARP Rat anti-Human, PerCP-eFluor 710, Clone: G14D9, eBioscience™

Rat Monoclonal Antibody

$201.55 - $520.55

Specifications

Antigen GARP
Clone G14D9
Host Species Rat
Gene Alias LRRC32, Garpin
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245948 Life Technologies
LS46988241
25 Tests Each for $201.55
50245949 Life Technologies
LS46988242
100 Tests Each for $520.55
Specifications

Description

The G14D9 monoclonal antibody reacts with human Glycoprotein A Repetitions Predominant (GARP, also known as LRRC32 or Garpin). GARP is an approximately 80kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on the surface of activated Treg cells has been reported to be necessary for their suppressive function, possibly related to its role as a cell surface receptor for LAP/TGF beta.
Description & Specifications

Specifications

GARP
Rat
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
G14D9
LRRC32, Garpin
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test