Please login to your online account to display your discounted pricing

GARP Rat anti-Human, PE-Cy7, Clone: G14D9, eBioscience™

Rat Monoclonal Antibody

$201.55 - $520.55

Specifications

Antigen GARP
Clone G14D9
Host Species Rat
Gene Alias LRRC32, Garpin
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123468

 
life technologies
LS25988241
25 tests Each for $201.55

501123469

 
life technologies
LS25988242
100 tests Each for $520.55
Description & Specifications

Specifications

Antigen GARP
Clone G14D9
Host Species Rat
Gene Alias LRRC32, Garpin
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1.0μg)/test

The G14D9 monoclonal antibody reacts with human Glycoprotein A Repetitions Predominant (GARP, also known as LRRC32 or Garpin). GARP is an approximately 80kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on the surface of activated Treg cells has been reported to be necessary for their suppressive function, possibly related to its role as a cell surface receptor for LAP/TGF beta.