Please login to your online account to display your discounted pricing

GAPDH Mouse anti-Human, Clone: FF26A, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $506.05

Specifications

Antigen GAPDH
Clone FF26A
Host Species Mouse
Species Reactivity Human
Applications Western Blotting
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122789

 
life technologies
LS14952380
25μg Each for $129.05

501122790

 
life technologies
LS14952382
100μg Each for $506.05
Description & Specifications

Specifications

Antigen GAPDH
Clone FF26A
Host Species Mouse
Species Reactivity Human
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL

This FF26A monoclonal antibody reacts with human glyceraldehyde-3-phosphate dehydrogenase (GAPDH), a key component of glycolysis. This 36kDa enzyme is constitutively expressed in all tissues albeit at different levels. Measurement of GAPDH levels is often performed as a control for protein loading in experiments assessing differential protein expression.

The FF26A antibody is crossreactive to mouse GAPDH.