Please login to your online account to display your discounted pricing

Alfa Aesar™ GAP 26

For cellular and molecular biology applications

Specifications

Product Type GAP 26
Molecular Weight 1550.87
Storage Requirements -30° to -10°C
Sequence Val-Cys-Tyr-Asp-Lys-Ser-Phe-Pro-Ile-Ser-His-Val-Arg
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64547EXD

 
alfa aesar™
J64547EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ64547MCR

 
alfa aesar™
J64547MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type GAP 26
Molecular Weight 1550.87
Storage Requirements -30° to -10°C
Sequence Val-Cys-Tyr-Asp-Lys-Ser-Phe-Pro-Ile-Ser-His-Val-Arg
Format Solid
Regulatory Status RUO
Purification Method HPLC