Please login to your online account to display your discounted pricing

GANT 61, Tocris

GLI antagonist; inhibits Hedgehog (Hh) signaling

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material GANT 61
Assay Percent Range >98%
Molecular Formula C27H35N5
Formula Weight 429.61
Synonym 2,2'-[[Dihydro-2-(4-pyridinyl)-1,3(2H,4H)-pyrimidinediyl]bis(methylene)]bis[N,N-dimethylbenzenamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

319110

 
r & d systems
319110
10mg Each for $208.25

319150

 
r & d systems
319150
50mg Each for $867.95
Description & Specifications

Specifications

Formula Weight 429.61
Quantity 10mg
Chemical Name or Material GANT 61
Assay Percent Range >98%
Recommended Storage Store at -20°C
Synonym 2,2'-[[Dihydro-2-(4-pyridinyl)-1,3(2H,4H)-pyrimidinediyl]bis(methylene)]bis[N,N-dimethylbenzenamine
Molecular Formula C27H35N5
CAS 500579-04-4
Safety and Handling

Store at -20°C