Please login to your online account to display your discounted pricing

Ganaxolone, Tocris

Potent, positive allosteric modulator of GABAA receptors

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Ganaxolone
Assay Percent Range >98%
Molecular Formula C22H36O2
Formula Weight 332.53
Synonym (3α, 5α)-3-Hydroxy-3-methyl-pregnan-20-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

253150

 
r & d systems
RDS253150
50mg Each for $746.48

253110

 
r & d systems
RDS253110
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 332.53
Quantity 50mg
Molecular Formula C22H36O2
Assay Percent Range >98%
Recommended Storage Store at +4°C
CAS 38398-32-2
Synonym (3α, 5α)-3-Hydroxy-3-methyl-pregnan-20-one
Chemical Name or Material Ganaxolone
Safety and Handling

Store at +4°C