Please login to your online account to display your discounted pricing

Alfa Aesar™ gamma-Undecanolactone, 99%

$55.11 - $132.53

Chemical Identifiers

Chemical Name or Material gamma-Undecanolactone
CAS 104-67-6
Molecular Formula C11H20O2
Formula Weight 184.28
MDL Number MFCD00005405
Beilstein 81943
Synonym 4-Hydroxyundecanoic acid lactone; gamma-Undecalactone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL16028AE

  SDS
alfa aesar™
L16028AE
100mL Each for $55.11

AAL16028AP

  SDS
alfa aesar™
L16028AP
500mL Each for $132.53
Description & Specifications

Specifications

Formula Weight 184.28
Chemical Name or Material gamma-Undecanolactone
Assay 99%
Molecular Formula C11H20O2
MDL Number MFCD00005405
CAS 104-67-6
Beilstein 81943
Synonym 4-Hydroxyundecanoic acid lactone; gamma-Undecalactone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 164° to 166°C (13mmHg)
RTECS Number YQ2485000
Quantity 100mL
Flash Point 156°C (312°F)
TSCA TSCA
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.