Please login to your online account to display your discounted pricing

gamma-Secretase Inhibitor XX

A dipeptidic (c)-secretase inhibitor

$885.97 - $885.97

Specifications

CAS 209984-56-5
Formula Weight 463.48g/mol
Synonym Deshydroxy LY 411575
Storage Requirements −30° to −10°C
Format Solid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ649040AM

 
alfa aesar™
J649040AM
Each for $885.97
Description & Specifications

Specifications

CAS 209984-56-5
Formula Weight 463.48g/mol
Synonym Deshydroxy LY 411575
Storage Requirements −30° to −10°C
Format Solid
For Use With (Application) A dipeptidic (c)-secretase inhibitor
Description White