missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

gamma-Oryzanol, TCI America™

Manufacturer:  TCI America O017225G

 View more versions of this product

Catalog No. O017225G



Specifications

Specifications

gamma-Oryzanol
25g
C40H58O4
2811
CC(CCC=C(C)C)C1CCC2(C1(CCC34C2CCC5C3(C4)CCC(C5(C)C)OC(=O)C=CC6=CC(=C(C=C6)O)OC)C)C
91971120
White-Yellow
11042-64-1
MFCD00867548
FODTZLFLDFKIQH-YGPNKWNYSA-N
602.9
Crystal-Powder at 20°C

Chemical Identifiers

11042-64-1
602.9
FODTZLFLDFKIQH-YGPNKWNYSA-N
CC(CCC=C(C)C)C1CCC2(C1(CCC34C2CCC5C3(C4)CCC(C5(C)C)OC(=O)C=CC6=CC(=C(C=C6)O)OC)C)C
C40H58O4
MFCD00867548
91971120
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0485

RTECSNumber : RN0050000

TSCA : No

Documents