missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ gamma-Neuropeptide, Rabbit

For cellular and molecular biology applications

$219.99 - $356.23

Specifications

Product Type gamma-Neuropeptide, rabbit
Cross Reactivity Rabbit
Molecular Weight 2320.61
Storage Requirements -30° to -10°C
Sequence Asp-Ala-Gly-His-Gly-Gln-Ile-Ser-His-Lys-Arg-His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66216EXD
View Documents
Alfa Aesar™
J66216EXD
0.5mg Each for $219.99
 
AAJ66216MCR
View Documents
Alfa Aesar™
J66216MCR
1mg Each for $356.23
 
Specifications

Specifications

gamma-Neuropeptide, rabbit
2320.61
Asp-Ala-Gly-His-Gly-Gln-Ile-Ser-His-Lys-Arg-His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2
γ-Preprotachykinin (72-92), amide
HPLC
Rabbit
-30° to -10°C
Solid
RUO

For Research Use Only