Please login to your online account to display your discounted pricing

Gamma-Linolenic acid, ≥99%, MP Biomedicals™


Quantity 1g

Manufacturer: mp biomedicals  0215524491

 View more versions of this product

Catalog No. ICN15524491

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material Gamma-Linolenic acid
CAS 506-26-3
Purity Grade ≥99%
Physical Form Clear colorless liquid
Molecular Formula C18H32O2
Formula Weight 278.4g/mol
Synonym 6, 9, 12-Octadecatrienoic acid

Specifications

Chemical Name or Material Gamma-Linolenic acid
CAS 506-26-3
Purity Grade ≥99%
Physical Form Clear colorless liquid
Molecular Formula C18H32O2
Formula Weight 278.4g/mol
Synonym 6, 9, 12-Octadecatrienoic acid