Please login to your online account to display your discounted pricing

Gamma-Linolenic acid, ≥99%, MP Biomedicals™

$1,019.92 - $1,019.92

Chemical Identifiers

Chemical Name or Material Gamma-Linolenic acid
CAS 506-26-3
Purity Grade ≥99%
Physical Form Clear colorless liquid
Molecular Formula C18H32O2
Formula Weight 278.4g/mol
Synonym 6, 9, 12-Octadecatrienoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15524491

 
mp biomedicals
0215524491
1g Each for $1,019.92
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Gamma-Linolenic acid
CAS 506-26-3
Purity Grade ≥99%
Physical Form Clear colorless liquid
Molecular Formula C18H32O2
Formula Weight 278.4g/mol
Synonym 6, 9, 12-Octadecatrienoic acid