Please login to your online account to display your discounted pricing

Alfa Aesar™ Gamma-L-Glutamyl Hydrazide


Manufacturer: alfa aesar™  J6767503

 View more versions of this product

Catalog No. AAJ6767503

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

CAS 1820-73-1
Molecular Formula C5H11N3O3
Formula Weight 161.16
Synonym (S)-2-amino-5-hydrazinyl-5-oxopentanoic acid; L-Glutamic acid γ-hydrazide
Sensitivity Light Sensitive
Quantity 1G
Melting Point 164-165° C
Solubility Soluble in water
Description White
Storage Requirements -30°C to -10°C
Format Powder
Type Gamma-L-Glutamyl Hydrazide
For Use With (Application) A modified amino acid; also known as H-L-Glu(NHNH2)-OH. A glutamine antagonist