Alfa Aesar™ Gamma-L-Glutamyl Hydrazide
Manufacturer:  Alfa Aesar™ J6767506
 View more versions of this product
Catalog No. AAJ6767506
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
5G
-30°C to -10°C
Gamma-L-Glutamyl Hydrazide
1820-73-1
161.16
Light Sensitive
Soluble in water
White
Powder
A modified amino acid; also known as H-L-Glu(NHNH2)-OH. A glutamine antagonist
C5H11N3O3
(S)-2-amino-5-hydrazinyl-5-oxopentanoic acid; L-Glutamic acid γ-hydrazide
164-165° C