Please login to your online account to display your discounted pricing

Alfa Aesar™ gamma-L-Glutamyl-4-nitroanilide, 98+%

$71.58 - $1,174.47

Chemical Identifiers

Chemical Name or Material gamma-L-Glutamyl-4-nitroanilide
CAS 7300-59-6
Molecular Formula C11H13N3O5
Formula Weight 267.24
MDL Number MFCD00036218
Beilstein 2818758
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1444203

  SDS
alfa aesar™
A1444203
1g Each for $71.58

AAA1444206

  SDS
alfa aesar™
A1444206
5g Each for $283.23

AAA1444214

  SDS
alfa aesar™
A1444214
25g Each for $1,174.47
Description & Specifications

Specifications

Formula Weight 267.24
Chemical Name or Material gamma-L-Glutamyl-4-nitroanilide
Melting Point 185° to 188°C
TSCA TSCA
Assay ≥98%
Molecular Formula C11H13N3O5
MDL Number MFCD00036218
CAS 7300-59-6
Beilstein 2818758
Quantity 1g