missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

gamma delta TCR Mouse anti-Human, PerCP-eFluor 710, Clone: B1.1, eBioscience™

Mouse Monoclonal Antibody

$154.44 - $389.40

Specifications

Antigen gamma delta TCR
Clone B1.1
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123983 Life Technologies
LS46995941
25 Tests Each for $154.44
501123984 Life Technologies
LS46995942
100 Tests Each for $389.40
Specifications

Description

The B1.1 monoclonal antibody reacts with the human gamma delta TCR complex. The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood; it is suggested that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulation of the immune response and during bacterial infection. This monoclonal can be used as a phenotypic marker for gamma delta TCR-expressing T cells.
Description & Specifications

Specifications

gamma delta TCR
Murine
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (2.0μg)/test
B1.1
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer