Please login to your online account to display your discounted pricing

gamma delta TCR Mouse anti-Human, PE, Clone: B1.1, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $333.50

Specifications

Antigen gamma delta TCR
Clone B1.1
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011322

 
life technologies
LS12995941
25 Tests Each for $143.55

5011323

 
life technologies
LS12995942
100 Tests Each for $333.50
Description & Specifications

Specifications

Antigen gamma delta TCR
Clone B1.1
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (4μg)/test

The B1.1 monoclonal antibody reacts with the human gamma delta TCR complex. The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood; it is suggested that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulation of the immune response and during bacterial infection. This monoclonal can be used as a phenotypic marker for gamma delta TCR-expressing T cells.